missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC25B Monoclonal Antibody (OTI11C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812342
This item is not returnable.
View return policy
Description
Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Directly dephosphorylates CDK1 and stimulates its kinase activity. The three isoforms seem to have a different level of activity.Specifications
| CDC25B | |
| Monoclonal | |
| Unconjugated | |
| CDC25B | |
| cell division cycle 25B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 994 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI11C5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30305 | |
| CDC25B | |
| Human recombit protein fragment corresponding to amino acids 421-567 of human CDC25B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction