missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDC25B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5111104

Product Code. 17169023

  • € 630.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LVKTLEKEEE KDLVMYSKCQ RLFRSPSMPC SVIRPILKRL ERPQDRDTPV QNKRRRSVTP PEEQQEAEEP KARVLRSKSL CHDEI Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 79%.

Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Directly dephosphorylates CDK1 and stimulates its kinase activity. The three isoforms seem to have a different level of activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDC25B
Polyclonal
Unconjugated
CDC25B
AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2
Rabbit
Antigen affinity chromatography
RUO
994
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P30305
CDC25B
Recombinant Protein corresonding to Human CDC25B. Recombinant protein control fragment (Product #RP-95267).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.