missing translation for 'onlineSavingsMsg'
Learn More

Cdc25C Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA585509

Product Code. 16342445

  • € 516.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Positive Control: Jurkat, Raji, K562, NCI-H929, HeLa (untreated), HeLa (100ng/ml Nocodazole treatment for 24hr), Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

M-phase inducer phosphatase 3is an enzyme that in humans is encoded by theCDC25Cgene. This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 (CDK1) and triggers entry into mitosis. Also, it is thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cdc25C
Polyclonal
Unconjugated
CDC25C
CDC25 antibody; CDC25C antibody; mitosis inducer CDC25 antibody; dual specificity phosphatase CDC25C antibody; M-phase inducer phosphatase 3 antibody; cell division cycle 25C antibody; protein phosphatase 1; regulatory subunit 60 antibody; phosphotyrosine phosphatase antibody; cell division cycle 25 homolog C (S. pombe) antibody
Rabbit
Antigen Affinity Chromatography
RUO
307511, 995, 12532
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
P48967, P30307
Cdc25c
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human CDC25C.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.