missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | CDC26 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699831
|
Novus Biologicals
NBP2-97346-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688894
|
Novus Biologicals
NBP2-97346-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC26 Polyclonal antibody specifically detects CDC26 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| CDC26 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.0) | |
| 246184 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ANAPC12chromosome 9 open reading frame 17, Anaphase-promoting complex subunit 12, anaphase-promoting complex subunit CDC26, APC12anaphase promoting complex subunit 12, C9orf17, CDC26 subunit of anaphase promoting complex, cell division cycle 26, cell division cycle 26 homolog (S. cerevisiae), Cell division cycle protein 26 homolog | |
| Produced in rabbits immunized with E. coli-derived Human CDC26 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title