missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc27 Antibody (AF 3.1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2606JF646
This item is not returnable.
View return policy
Description
Cdc27 Monoclonal antibody specifically detects Cdc27 in Mammalia, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOF
Specifications
| Cdc27 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog | |
| A peptide corresponding to the C-terminal 10 residues of human Cdc27 coupled to KLH. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 996 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| AF 3.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mammalia, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction