missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC2L2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-495B
This item is not returnable.
View return policy
Description
CDC2L2 Polyclonal antibody specifically detects CDC2L2 in Human samples. It is validated for Western Blot
Specifications
| CDC2L2 | |
| Polyclonal | |
| Western Blot | |
| CDK11-p110, CDK11-p58, cyclin-dependent kinase 11A, PITSLRE protein kinase beta | |
| The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human Cell Division Cycle 2-like 2 using the numbering given in SwissProt entry Q9UQ88 (GeneID 985). | |
| 0.1 mL | |
| Apoptosis, Cell Cycle and Replication, Cellular Markers, Protein Phosphatase, Signal Transduction | |
| 728642 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction