missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC2L5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34123
This item is not returnable.
View return policy
Description
CDC2L5 Polyclonal specifically detects CDC2L5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| CDC2L5 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Q14004 | |
| CDK13 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PLGGIQPSSQTIQPKVETDAAQAAVQSAFAVLLTQLIKAQQSKQKDVLLEERENGSGHEASLQLRPPPEPSTPVSGQDDLIQHQDMRILELTPEPDRPRILPPDQRPPE | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8621 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CDC2L5, CDC2LEC 2.7.11.22, CDC2-related protein kinase 5, Cell division cycle 2-like protein kinase 5, Cell division protein kinase 13, CHEDcell division cycle 2-like 5 (cholinesterase-related cell division controller), Cholinesterase-related cell division controller, cyclin-dependent kinase 13, EC 2.7.11, EC 2.7.11.23, hCDK13, KIAA1791FLJ35215 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction