missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37 Antibody (3G7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59466G
This item is not returnable.
View return policy
Description
CDC37 Monoclonal antibody specifically detects CDC37 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| CDC37 | |
| Monoclonal | |
| DyLight 488 | |
| CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
| Recombinant human CDC37 (1-378aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
| 11140 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 3G7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction