missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99422-100ul
This item is not returnable.
View return policy
Description
CDC37 Polyclonal antibody specifically detects CDC37 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| CDC37 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 4-8 uL/mg of lysate | |
| CDC37 (cell division cycle 37, S. cerevisiae, homolog), CDC37 cell division cycle 37 homolog, CDC37 cell division cycle 37 homolog (S. cerevisiae), CDC37A, cell division cycle 37 homolog (S. cerevisiae), Hsp90 chaperone protein kinase-targeting subunit, hsp90 co-chaperone Cdc37, P50CDC37 | |
| Produced in rabbits immunized with purified, recombinant Human CDC37 (Accession#: NP_008996.1; Met1-Val378) | |
| 100 μg | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
| 11140 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction