missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Cdc37 (Ser13) Recombinant Rabbit Monoclonal Antibody (5P8B2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA535890

Product Code. 17136543

  • € 548.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EVSDD.

In S. cerevisiae, Cdc37 has been shown to be an important regulator of cell division cycle through its action on cyclin-dependent kinases (CDK's). Cdc37 has also been associated with signaling pathways and kinases unrelated to cell cycle control, including v-Src, casein kinase II, MPS1 kinase and the sevenless receptor tyrosine kinase. Interestingly, HSP90 has also been shown to be associated with many of the same cellular targets as Cdc37. It has now been shown that Cdc37 is the same protein previously described as p50 which has been found to be associated with many of the same proteins as HSP90. Among others, Cdc37/p50 has been found complexed with Src-family kinases (Fyn, Yes, Fes, Lck & Fgr), Raf and pp60v-src. More recently, Cdc37 has been shown to be a chaperone independently and in concert with HSP90. In fact, Cdc37 is necessary for the binding of HSP90 to Cdk4.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-Cdc37 (Ser13)
Recombinant Monoclonal
0.09 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q16543, Q61081, Q63692
CDC37
A synthetic phosphorylated peptide around S13 of human CDC37 (Q16543).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
5P8B2
Unconjugated
CDC37
CDC 37; CDC37; CDC37 (cell division cycle 37 S. cerevisiae homolog); CDC37 (cell division cycle 37, S. cerevisiae, homolog); CDC37 cell division cycle 37 homolog; Cdc37 homolog; CDC37 protein; CDC37A; Cdc37p; Cell division control protein 37; cell division cycle 37; cell division cycle 37 homolog; hsp90 chaperone protein kinase-targeting subunit; Hsp90 co-chaperone Cdc37; Hsp90 co-chaperone Cdc37, N-terminally processed; p50; P50CDC37; SMO1; YD9489.03; YDR168W
Rabbit
Affinity Chromatography
RUO
11140, 114562, 12539
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.