missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC37L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98987-50ul
This item is not returnable.
View return policy
Description
CDC37L1 Polyclonal antibody specifically detects CDC37L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CDC37L1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| CDC37 cell division cycle 37 homolog (S. cerevisiae)-like 1, CDC37BHARCCDC37 cell division cycle 37 homolog-like 1, cell division cycle 37 homolog (S. cerevisiae)-like 1, FLJ20639, hsp90 co-chaperone Cdc37-like 1, Hsp90-associating relative of Cdc37 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CDC37L1. | |
| 50 μg | |
| Cell Cycle and Replication | |
| 55664 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction