missing translation for 'onlineSavingsMsg'
Learn More

CDC37L1 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16855884 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16855884 150 μL 150µL
16865884 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16855884 Supplier Proteintech Supplier No. 162931AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 ).

Specifications

Antigen CDC37L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDC37L1
Gene Accession No. Q7L3B6
Gene Alias CDC37B, CDC37L1, HARC
Gene Symbols CDC37L1
Host Species Rabbit
Immunogen CDC37L1 Fusion Protein Ag9459
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55664
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.