missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 ).
Specifications
Specifications
| Antigen | CDC37L1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.17 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CDC37L1 |
| Gene Accession No. | Q7L3B6 |
| Gene Alias | CDC37B, CDC37L1, HARC |
| Gene Symbols | CDC37L1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?