missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc40 Recombinant Rabbit Monoclonal Antibody (23GB3445), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553382
This item is not returnable.
View return policy
Description
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is found to be essential for the catalytic step II in pre-mRNA splicing process. It is found in the spliceosome, and contains seven WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp17 protein, which functions in two different cellular processes: pre-mRNA splicing and cell cycle progression. It suggests that this protein may play a role in cell cycle progression.
Specifications
| Cdc40 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CDC40 | |
| 1200003H23Rik; Cdc40; cell division cycle 40; cell division cycle 40 homolog; EHB3; EH-binding protein 3; hPRP17; pre-mRNA splicing factor 17; pre-mRNA-processing factor 17; PRP17; PRP17 homolog; PRPF17 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361859, 51362, 71713 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3445 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60508, Q9DC48 | |
| CDC40 | |
| A synthetic peptide of human CDC40 (50-100AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction