Learn More
CDC42 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98725APC
Product Code. 18601712
  • € 430.00 / 0.10mL
Description
CDC42 Polyclonal antibody specifically detects CDC42 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunoprecipitation
APC
0.1 mL
Breast Cancer, Cancer, Phospho Specific, Signal Transduction
Polyclonal
Antigen and protein A Affinity-purified