missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-76330JF549
This item is not returnable.
View return policy
Description
CDC42 Polyclonal antibody specifically detects CDC42 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| CDC42 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Janelia Fluor 549 | |
| CDC42Hs, cell division control protein 42 homolog, cell division cycle 42 (GTP binding protein, 25kDa), cell division cycle 42 (GTP-binding protein, 25kD), dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)), dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)), G25K, G25K GTP-binding protein, growth-regulating protein, GTP-binding protein, 25kD, small GTP binding protein CDC42 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human CDC42. The immunogen is located within amino acids 40 - 90 of CDC42. Amino Acid Squence: FDTAGQEDYDRLRPLSY | |
| 0.1 mL | |
| Breast Cancer, Cancer, Signal Transduction | |
| 998 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction