missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDC42 BPG (MRCKG), GST Tag Recombinant Protein

Product Code. 17168725
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 17168725

Brand: Invitrogen™ A32861

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDC42 BPG (MRCKG) recombinant human protein (1-475) was expressed in insect cells. CDC42 BPG (MRCKG) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for CDC42 BPG (MRCKG) is NP_059995.2.

CDC42 BPG serine/threonine kinase is encoded by the CDC42BPG gene. It acts as a downstream effector of CDC42 in cytoskeletal reorganization, and contributes to the actomyosin contractility required for cell invasion, through the regulation of MYPT1 and MLC2 phosphorylation. It belongs to the AGC protein kinase family, DMPK subfamily. It contains an N-terminal kinase domain, followed by a coiled-coil domain, a second coiled-coil domain containing a kinase inhibitory motif (KIM), a variable region, a cysteine-rich domain, a pleckstrin (PLEK; 173570) homology (PH) domain, a citron (605629) homology (CNH) domain, and a C-terminal CDC42 (116952)/RAC (602048) interactive binding (CRIB) domain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6DT37
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 0.25mM DTT, 0.1mM EDTA, 150mM NaCl, 10mM glutathione, 0.1mM PMSF and no preservative; pH 7.5
Gene ID (Entrez) 55561
Molecular Weight (g/mol) 80.2 kDa
Name Human CDC42 BPG (MRCKG), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BC046418; CDC42 binding protein kinase gamma; CDC42 binding protein kinase gamma (DMPK-like); CDC42-binding protein kinase gamma; CDC42BPG; DMPK2; DMPK-like gamma; HSMDPKIN; KAPPA-200; MRCK gamma; MRCKG; MRCKgamma; Myotonic dystrophy kinase-related CDC42-binding kinase gamma; myotonic dystrophy protein kinase like protein; myotonic dystrophy protein kinase-like alpha; myotonic dystrophy protein kinase-like gamma; serine/threonine-protein kinase MRCK gamma
Common Name MRCK gamma
Gene Symbol CDC42BPG
KinaseFamily Other Kinase Family
Protein Length 1-475
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-475
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.