missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42BPA Monoclonal Antibody (OTI7B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808223
This item is not returnable.
View return policy
Description
MRCK alpha (myotonic dystrophy kinase-related Cdc42-binding kinase alpha) is a member of the dystrophia myotonica protein kinase (DMPK) family that functions downstream of Cdc42 in actin cytoskeletal reorganization. It is a serine/threonine kinase with multiple functional domains, including phorbol ester-responsive C1 domain. Three independent coiled-coil domains and the N-terminal region preceding the kinase domain are responsible for intermolecular interactions leading to MRCK alpha multimerization. The transautophosphorylation process is critical for regulation of MRCK alpha catalytic activities. Binding of phorbol esters to MRCK releases its autoinhibition, allowing N-terminal dimerization and subsequent kinase activation.Specifications
| CDC42BPA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5VT25 | |
| CDC42BPA | |
| Human recombit protein fragment corresponding to amino acids 1419-1638 of human CDC42BPA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7B9 | |
| Unconjugated | |
| CDC42BPA | |
| MRCK, MRCKA, PK428 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8476 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction