missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42BPB Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98656R
This item is not returnable.
View return policy
Description
CDC42BPB Polyclonal specifically detects CDC42BPB in Human samples. It is validated for Western Blot.
Specifications
| CDC42BPB | |
| Polyclonal | |
| Western Blot | |
| CDC42 binding protein kinase beta (DMPK-like), CDC42-binding protein kinase beta, CDC42-binding protein kinase beta (DMPK-like), DMPK-like beta, EC 2.7.11, EC 2.7.11.1, FLJ37601, FLJ44730, KIAA1124DKFZp686H1431, MRCK beta, MRCKB, Myotonic dystrophy kinase-related CDC42-binding kinase beta, Myotonic dystrophy protein kinase-like beta, serine/threonine-protein kinase MRCK beta | |
| Produced in rabbits immunized with E. coli-derived Human CDC42BPB fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 9578 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction