missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42BPB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | CDC42BPB |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620582
|
Novus Biologicals
NBP2-98656-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694395
|
Novus Biologicals
NBP2-98656-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC42BPB Polyclonal antibody specifically detects CDC42BPB in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| CDC42BPB | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.0) | |
| 9578 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CDC42 binding protein kinase beta (DMPK-like), CDC42-binding protein kinase beta, CDC42-binding protein kinase beta (DMPK-like), DMPK-like beta, EC 2.7.11, EC 2.7.11.1, FLJ37601, FLJ44730, KIAA1124DKFZp686H1431, MRCK beta, MRCKB, Myotonic dystrophy kinase-related CDC42-binding kinase beta, Myotonic dystrophy protein kinase-like beta, serine/threonine-protein kinase MRCK beta | |
| Produced in rabbits immunized with E. coli-derived Human CDC42BPB fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title