missing translation for 'onlineSavingsMsg'
Learn More

CDC42EP3 Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30516272 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30516272 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30516272 Supplier Novus Biologicals Supplier No. NBP338553PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDC42EP3 Polyclonal antibody specifically detects CDC42EP3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC42EP3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias Binder of Rho GTPases 2, BORG2CEP3UB1, CDC42 effector protein (Rho GTPase binding) 3, cdc42 effector protein 3, CRIB-containing BORG2 protein, FLJ46903, MSE55-related Cdc42-binding protein, MSE55-related protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-254 of human CDC42EP3 (NP_006440.2).,, Sequence:, MPAKTPIYLKAANNKKGKKFKLRDILSPDMISPPLGDFRHTIHIGKEGQHDVFGDISFLQGNYELLPGNQEKAHLGQFPGHNEFFRANSTSDSVFTETPSPVLKNAISLPTIGGSQALMLPLLSPVTFNSKQESFGPAKLPRLSCEPVMEEKAQEKSSLLENGTVHQGDTSWGSSGSASQSSQGRDSHSSSLSEQYPDWPAEDMFDHPTPCELIKGKTKSEESLSDLTGSLLSLQLDLGPSLLDEVLNVMDKNK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10602
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.