missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42SE2 Monoclonal Antibody (OTI10A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808200
This item is not returnable.
View return policy
Description
Probably involved in the organization of the actin cytoskeleton by acting downstream of CDC42, inducing actin filament assembly. Alters CDC42-induced cell shape changes. In activated T-cells, may play a role in CDC42-mediated F-actin accumulation at the immunological synapse. May play a role in early contractile events in phagocytosis in macrophages.Specifications
| CDC42SE2 | |
| Monoclonal | |
| Unconjugated | |
| CDC42SE2 | |
| SPEC2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56990 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI10A5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NRR3 | |
| CDC42SE2 | |
| Full length human recombit protein of human CDC42SE2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction