Learn More
CDC42SE2 Monoclonal Antibody (OTI10A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808200S
Encompass_Preferred
 View more versions of this product
Product Code. 17519193
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
CDC42SE2 Monoclonal antibody specifically detects CDC42SE2 in Human samples. It is validated for Western Blot
Specifications
Specifications
CDC42SE2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O15371
Mouse
Affinity chromatography
RUO
8664
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI10A5
Unconjugated
EIF3D
2810404F18Rik; AA408783; AA536669; CDC42 small effector 2; CDC42 small effector protein 2; Cdc42se2; non-kinase Cdc42 effector protein SPEC2; RGD1563924; small effector of CDC42 protein 2; SPEC2
Full length human recombinant protein of human EIF3D produced in HEK293T cell.
30 μL