missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC5L Antibody (PCRP-CDC5L-2C6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24018AF647
This item is not returnable.
View return policy
Description
CDC5L Monoclonal antibody specifically detects CDC5L in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| CDC5L | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CDC5, CDC5 (cell division cycle 5, S. pombe, homolog)-like, CDC5 cell division cycle 5-like (S. pombe), Cdc5-like protein, Cdc5-related protein, CEF1, cell division cycle 5-like protein, dJ319D22.1, dJ319D22.1 (CDC5-like protein), hCDC5, KIAA0432, PCDC5RPCDC5-LIKE, Pombe cdc5-related protein | |
| Recombinant fragment (around aa 1-148) of human CDC5L protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-CDC5L-2C6 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 988 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction