missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc6 Antibody (OTI1A4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70383UV
This item is not returnable.
View return policy
Description
Cdc6 Monoclonal specifically detects Cdc6 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cdc6 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDC6 | |
| Full length recombinant protein of human Cdc6 (NP_001245) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1A4 | |
| Western Blot, Immunohistochemistry | |
| CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6) | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| 990.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction