missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC6 Monoclonal Antibody (OTI7B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808380S
This item is not returnable.
View return policy
Description
CDC6 Monoclonal antibody specifically detects CDC6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CDC6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60393 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 135935 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7B12 | |
| Unconjugated | |
| NOBOX | |
| CDC18(S.pombe); CDC18L; cdc18-related protein; CDC6; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6; cell division cycle 6 homolog; HsCDC18; HsCDC6; p62(cdc6) | |
| Human recombinant protein fragment corresponding to amino acids 1-271 of human NOBOX produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction