missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC6 Monoclonal Antibody (OTI8E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808458
This item is not returnable.
View return policy
Description
The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins.Specifications
| CDC6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99741 | |
| CDC6 | |
| Full length human recombit protein of human CDC6 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI8E6 | |
| Unconjugated | |
| CDC6 | |
| CDC18L, HsCDC18, HsCDC6, MGORS5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 990 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction