Learn More
CDC6 Monoclonal Antibody (OTI8E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808458S
Encompass_Preferred
 View more versions of this product
Product Code. 17569743
  • € 242.00 / 30µL
Description
CDC6 Monoclonal antibody specifically detects CDC6 in Human samples. It is validated for Western Blot
Specifications
Specifications
CDC6
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96GD3
Mouse
Affinity chromatography
RUO
22955
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI8E6
Unconjugated
SCMH1
CDC18(S.pombe); CDC18L; cdc18-related protein; CDC6; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6; cell division cycle 6 homolog; HsCDC18; HsCDC6; p62(cdc6)
Human recombinant protein fragment corresponding to amino acids 1-309 of human SCMH1 produced in E.coli.
30 μL