missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC7 Antibody (DCS-341) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 232.00
Specifications
| Antigen | CDC7 |
|---|---|
| Clone | DCS-341 |
| Concentration | 2 mg/mL |
| Dilution | Western Blot 0.2 - 0.4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Microarray |
| Applications | Western Blot |
Description
CDC7 Monoclonal specifically detects CDC7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray.Specifications
| CDC7 | |
| 2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, GPCR, Stem Cell Markers | |
| CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1, CDC7 cell division cycle 7 (S. cerevisiae), CDC7L1MGC117361, CDC7-related kinase, cell division cycle 7 (S. cerevisiae), cell division cycle 7 homolog (S. cerevisiae), cell division cycle 7-like protein 1, cell division cycle 7-related protein kinase, EC 2.7.11.1, HsCdc7, Hsk1, huCdc7, MGC126237, MGC126238 | |
| CDC7 | |
| IgG1 | |
| Protein A or G purified | |
| Mouse Monoclonal to Cdc7 Kinase (DCS-341) |
| DCS-341 | |
| Western Blot 0.2 - 0.4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500, Microarray | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 8317 | |
| full-length human Cdc7 kinase. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 55 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title