missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CDC7 Antibody (SR2732), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 316.00 - € 382.00
Specifications
| Antigen | CDC7 |
|---|---|
| Clone | SR2732 |
| Dilution | Western Blot 1:1000-1:2000, Flow Cytometry 1:100 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041649
|
Novus Biologicals™
NBP3-27710-100ul |
100 μL |
€ 382.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041710
|
Novus Biologicals™
NBP3-27710-50ul |
50 μL |
€ 316.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDC7 Monoclonal antibody specifically detects CDC7 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western BlotSpecifications
| CDC7 | |
| Western Blot 1:1000-1:2000, Flow Cytometry 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1, CDC7 cell division cycle 7 (S. cerevisiae), CDC7L1MGC117361, CDC7-related kinase, cell division cycle 7 (S. cerevisiae), cell division cycle 7 homolog (S. cerevisiae), cell division cycle 7-like protein 1, cell division cycle 7-related protein kinase, EC 2.7.11.1, HsCdc7, Hsk1, huCdc7, MGC126237, MGC126238 | |
| A synthesized peptide derived from human CDC7 (Uniprot #: O00311) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR2732 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 8317 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title