missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDCA4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16327214
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16327214 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16327214 Supplier Invitrogen™ Supplier No. PA578517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: NCI-H929 Predicted Reactivity: Chimpanzee (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CDCA4 encodes a protein that belongs to the E2F family of transcription factors. This protein regulates E2F-dependent transcriptional activation and cell proliferation, mainly through the E2F/retinoblastoma protein pathway. It also functions in the regulation of JUN oncogene expression. This protein shows distinctive nuclear-mitotic apparatus distribution, it is involved in spindle organization from prometaphase, and may also play a role as a midzone factor involved in chromosome segregation or cytokinesis. Two alternatively spliced transcript variants encoding the same protein have been noted for this gene. Two pseudogenes have also been identified on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene CDCA4
Gene Accession No. Q9BXL8
Gene Alias 2410018C03Rik; Cdca4; cell division cycle associated 4; cell division cycle-associated protein 4; fb78b08; FLJ20764; Hematopoietic progenitor protein; HEPP; hypothetical protein LOC494037; MGC19517; SEI-3; SEI-3/HEPP; si:dkey-177p2.7; wu:fb78b08; zgc:92217
Gene Symbols CDCA4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CDCA4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55038
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.