missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDCA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109502
This item is not returnable.
View return policy
Description
Immunogen sequence: VPAVQSPRRS PRISFFLEKE NEPPGRELTK EDLFKTHSVP ATPTSTPVPN PEAESSSKEG ELDARDLEMS KKVRRSYSRL E.
CDCA5 is the regulator of sister chromatid cohesion in mitosis. It may act by regulating the ability of the cohesin complex to mediate sister chromatid cohesion, perhaps by altering the nature of the interaction of cohesin with the chromosomes.
Specifications
| CDCA5 | |
| Polyclonal | |
| Unconjugated | |
| CDCA5 | |
| 2610036L13Rik; AL024086; AW536684; C85404; Cdca5; cell division cycle associated 5; cell division cycle-associated protein 5; p35; RGD1560863; sororin; sororin p35 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113130, 67849, 684771 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.81 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96FF9, Q9CPY3 | |
| CDCA5 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction