missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDCA5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15526682
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15526682 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15526682 Supplier Invitrogen™ Supplier No. PA544877

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: ATPTSTPVPN PEAESSSKEG ELDARDLEMS KKVRRSYSRL ETLGSASTST Sequence homology: Human: 100%.

CDCA5 is the regulator of sister chromatid cohesion in mitosis. It may act by regulating the ability of the cohesin complex to mediate sister chromatid cohesion, perhaps by altering the nature of the interaction of cohesin with the chromosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDCA5
Gene Accession No. Q96FF9
Gene Alias 2610036L13Rik; AL024086; AW536684; C85404; Cdca5; cell division cycle associated 5; cell division cycle-associated protein 5; p35; RGD1560863; sororin; sororin p35
Gene Symbols CDCA5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CDCA5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113130
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.