missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA7L Antibody (OTI3B5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72009AF700
This item is not returnable.
View return policy
Description
CDCA7L Monoclonal specifically detects CDCA7L in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CDCA7L | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CDCA7L | |
| Recombinant protein fragment corresponding to amino acids 1-337 of human CDCA7L (NP_061189) produced in E.coli. | |
| 0.1 mL | |
| 55536.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3B5 | |
| Western Blot, Immunohistochemistry | |
| cell division cycle associated 7-like, DKFZp762L0311, JPO2cell division cycle-associated 7-like protein, Protein JPO2, R1HR1, RAM2, Transcription factor RAM2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction