missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA7L Mouse anti-Human, Clone: OTI3B5, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803020S
This item is not returnable.
View return policy
Description
CDCA7L Monoclonal antibody specifically detects CDCA7L in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CDCA7L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96GN5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55536 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B5 | |
| Unconjugated | |
| CDCA7L | |
| BC006933; CDCA7L; cell division cycle associated 7 like; cell division cycle associated 7-like; cell division cycle-associated 7-like protein; HR1; JPO2; Protein JPO2; R1; RAM2; transcription factor RAM2 | |
| Human recombinant protein fragment corresponding to amino acids 1-337 of human CDCA7L produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction