missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDCA7L Polyclonal Antibody
GREENER_CHOICE

Product Code. 16609844
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16609844 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16609844 Supplier Invitrogen™ Supplier No. PA5103389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CDCA7L.

Cell division cycle-associated 7-like protein (or RAM2) is a transcription factor which plays a role in transcriptional regulation and apoptotic signaling pathways. It acts as a repressor that inhibits monoamine oxidase A (MAOA) activity and gene expression by binding to promoter. It also plays an important oncogenic role in mediating the full transforming effect of MYC in medulloblastoma cells. It may act downstream of P38-kinase and BCL-2, but upstream of CASP3/caspase-3 as well as CCND1/cyclin D1 and E2F1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA7L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDCA7L
Gene Accession No. Q96GN5
Gene Alias BC006933; CDCA7L; cell division cycle associated 7 like; cell division cycle associated 7-like; cell division cycle-associated 7-like protein; HR1; JPO2; Protein JPO2; R1; RAM2; transcription factor RAM2
Gene Symbols CDCA7L
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDCA7L(Accession Q96GN5), corresponding to amino acid residues M219-M269.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55536
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.