missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCP1 Antibody (309121), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB26662H
This item is not returnable.
View return policy
Description
CDCP1 Monoclonal antibody specifically detects CDCP1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF
Specifications
| CDCP1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, CyTOF | |
| 309121 | |
| Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
| CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
| Mouse myeloma cell line NS0-derived recombinant human CDCP1 isoform 1, Ala33-Leu666, Accession # NP_073753 | |
| 0.1 mL | |
| Phospho Specific | |
| 64866 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?