missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH1 Monoclonal Antibody (OTI3C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803253S
This item is not returnable.
View return policy
Description
CDH1 Monoclonal antibody specifically detects CDH1 in Human samples. It is validated for Western BlotSpecifications
| CDH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43699 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7080 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3C11 | |
| Unconjugated | |
| NKX2-1 | |
| AA960649; ARC-1; cadherin 1; cadherin 1, E-cadherin (epithelial); cadherin 1, epithelial; cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin e; cadherin-1; cadherin-E; calcium-dependent adhesion protein, epithelial; CAM 120/80; CD324; CDH1; CDHE; cell adhesion molecule; cell-CAM 120/80; ECAD; E-cad; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; Ecadherin; E-cadherin; E-cadherin 1; E-cadherin precursor; Epithelial cadherin; hab; half baked; LCAM; L-CAM; Um; UVO; Uvomorulin | |
| Full length human recombinant protein of human NKX2-1 ( NP_003308) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction