Learn More
CDH1 Monoclonal Antibody (OTI4F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800692
Encompass_Preferred
 View more versions of this product
Product Code. 17598772
  • € 540.87 / 100µL
Description
E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintece of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.
Specifications
Specifications
CDH1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P12830
CDH1
Full length human recombit protein of human CDH1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI4F1
Unconjugated
CDH1
Arc-1, BCDS1, CD324, CDHE, ECAD, LCAM, UVO
Mouse
Affinity Chromatography
RUO
999
-20° C, Avoid Freeze/Thaw Cycles