Learn More
CDH22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA538857
Product Code. 15535771
  • € 484.10 / 100µg
Description
This gene is a member of the cadherin superfamily. The gene product is composed of five cadherin repeat domains and a cytoplasmic tail similar to the highly conserved cytoplasmic region of classical cadherins. Expressed predomitly in the brain, this putative calcium-dependent cell adhesion protein may play an important role in morphogenesis and tissue formation in neural and non-neural cells during development and maintece of the brain and neuroendocrine organs. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDH22
Polyclonal
Unconjugated
CDH22
C20orf25
Rabbit
Antigen affinity chromatography
RUO
104010, 29182, 64405
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UJ99, Q9WTP5, Q63315
CDH22
A synthetic peptide derived from the internal region of human CDH22