missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH23 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519000
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
This gene is a member of the cadherin superfamily, whose genes encode calcium dependent cell-cell adhesion glycoproteins. The encoded protein is thought to be involved in stereocilia organization and hair bundle formation. The gene is located in a region containing the human deafness loci DFNB12 and USH1D. Usher syndrome 1D and nonsyndromic autosomal recessive deafness DFNB12 are caused by allelic mutations of this cadherin-like gene. Alternative splice variants encoding different isoforms have been described.
Specifications
| CDH23 | |
| Polyclonal | |
| Unconjugated | |
| CDH23 | |
| 4930542A03Rik; age related hearing loss 1; ahl; ahl1; bob; bobby; bus; bustling; Cadherin; cadherin 23; cadherin 23 (otocadherin); cadherin related 23; cadherin-23; cadherin-like 23; cadherin-related 23; cadherin-related family member 23; Cdh23; CDHR23; DFNB12; DKFZp434P2350; FLJ00233; FLJ36499; KIAA1774; KIAA1812; mdfw; MGC102761; modifier of deaf waddler; nmf112; nmf181; nmf252; otocadherin; OTTHUMP00000044780; sals; UNQ1894/PRO4340; USH1D; USH1H; v; W; waltzer; Waltzing | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 64072 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H251 | |
| CDH23 | |
| Synthetic peptide sequence (YNISLYENVTVGTS ) corresponding to the internal amino acids of CDH23 (aa 465-478). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction