Learn More
CCT4 Monoclonal Antibody (OTI1A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810896
This item is not returnable.
View return policy
Description
The chaperonin containing TCP1 complex (CCT), also called the TCP1 ring complex, consists of 2 back-to-back rings, each containing 8 unique but homologous subunits, such as CCT4. CCT assists the folding of newly translated polypeptide substrates through multiple rounds of ATP-driven release and rebinding of partially folded intermediate forms. Substrates of CCT include the cytoskeletal proteins actin and tubulin, as well as alpha-transducin.Specifications
| CCT4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P50991 | |
| CCT4 | |
| Full length human recombit protein of human CCT4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1A3 | |
| Unconjugated | |
| CCT4 | |
| CCT-DELTA, Cctd, SRB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10575 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |