Learn More
CDK1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10762-100UL
Description
The CDK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CDK1. This antibody recognizes human antigen. The CDK1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| CDK1 | |
| 1000 μg/mL | |
| CDC2, CDC28A, CDK1, Cell division protein kinase 1, Cyclin dependent kinase 1, DKFZp686L20222, p34 protein kinase | |
| 100 μL | |
| 983 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |