Learn More
CDK1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-10762-100UL
 View more versions of this product
Product Code. 18026175
  • € 450.00 / 100µL
Description
The CDK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CDK1. This antibody recognizes human antigen. The CDK1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
CDK1
1000 μg/mL
CDC2, CDC28A, CDK1, Cell division protein kinase 1, Cyclin dependent kinase 1, DKFZp686L20222, p34 protein kinase
100 μL
983
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa