missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584741
This item is not returnable.
View return policy
Description
Immunogen sequence: ALKKVRMDKE KDGIPISSLR EITLLLRLRH PNIVELKEVV VGNHLESIFL VMGYCEQDLA SLL Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene belongs to the CDK subfamily of the Ser/Thr protein kinase family. The CDK subfamily members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and are known to be essential for cell cycle progression. This kinase has been shown to play a role in cellular proliferation and its function is limited to cell cycle G2-M phase. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| CDK10 | |
| Polyclonal | |
| Unconjugated | |
| CDK10 | |
| BC017131; CDC2-related protein kinase; Cdk10; Cell division protein kinase 10; cyclin dependent kinase 10; cyclin-dependent kinase (CDC2-like) 10; cyclin-dependent kinase 10; cyclin-dependent kinase related protein; PISSLRE; serine/threonine protein kinase PISSLRE; serine/threonine-protein kinase PISSLRE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8558 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15131 | |
| CDK10 | |
| Recombinant protein corresponding to Human CDK10. Recombinant protein control fragment (Product #RP-107540). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction