missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK11A/CDK11B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538829
This item is not returnable.
View return policy
Description
The PITSLRE beta1 protein, a distantly related member of the Cdk family of protein kinases, induces apoptosis after low levels of ectopic expression. Apoptosis, or programmed cell death, is similarly induced by ectopic expression of an amino terminal deletion mutant retaining the catalytic and carboxyterminal domains of PITSLRE beta1, but not by other mutants lacking Histone H1 kinase activity or by other Cdk family members. The terminology for the ten isoforms of the PITSLRE subfamily of proteins is based on the conserved PSTAIRE box region of Cdc2 p34. Depending on which of the PITSLRE genes produce the protein, the cDNA and protein are designated alpha, beta or gamma (i.e., PITSLRE A gene, alpha; PITSLRE B gene, beta and PITSLRE C gene, gamma). Some of the isoforms such as PITSLRE alpha1 (T cells) and PITSLRE beta1 (B cells and brain), are expressed in specific cell types, while others are expressed ubiquitously.
Specifications
| CDK11A/CDK11B | |
| Polyclonal | |
| Unconjugated | |
| CDK11A | |
| CDC2L2, CDC2L3, CDK11, CDK11 p110, CDK11 p46, CDK11 p58, CDK11A, CDK8 like cyclin dependent kinase, Cell division cycle 2 like 2 (PITSLRE proteins), Cell division cycle 2 like protein kinase 2, Cell division cycle 2-like protein kinase 2, Cell division protein kinase 11A, Cyclin dependent kinase (CDC2 like) 11, Cyclin-dependent kinase 11A, Death preventing kinase, Galactosyltransferase associated protein kinase p58/GTA, Galactosyltransferase-associated protein kinase p58/GTA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 728642, 984 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P21127, Q9UQ88 | |
| CDK11A, CDK11B | |
| A synthetic peptide derived from the internal region of human CDK11 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction