missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK2 Antibody (AN4.3), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2721MFV500
This item is not returnable.
View return policy
Description
CDK2 Monoclonal antibody specifically detects CDK2 in Human, Mouse, Xenopus samples. It is validated for Flow Cytometry
Specifications
| CDK2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Xenopus species | |
| Purified |
| Flow Cytometry | |
| AN4.3 | |
| Flow Cytometry | |
| cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2) | |
| Human recombinant CDK2 | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| 1017 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction