missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 236.00 - € 541.00
Specifications
| Antigen | CDK2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227886
|
Novus Biologicals
NBP3-33184-20ul |
20 μL |
€ 236.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230226
|
Novus Biologicals
NBP3-33184-100ul |
100 μL |
€ 541.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDK2 Monoclonal antibody specifically detects CDK2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| CDK2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 1017 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2) | |
| A synthetic peptide corresponding to a sequence within amino acids 199-298 of human CDK2 (P24941).,, Sequence:, RRALFPGDSEIDQLFRIFRTLGTPDEVVWPGVTSMPDYKPSFPKWARQDFSKVVPPLDEDGRSLLSQMLHYDPNKRISAKAALAHPFFQDVTKPVPHLRL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title