missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK2 Mouse anti-Human, Clone: OTI1C4, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502807S
This item is not returnable.
View return policy
Description
CDK2 Monoclonal antibody specifically detects CDK2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CDK2 | |
| Monoclonal | |
| 0.65 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P24941 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1017 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1C4 | |
| Unconjugated | |
| CDK2 | |
| A630093N05Rik; CDC28; CDC2A; cdc2-related protein kinase; CDK1; CDK2; CDKN2; Cell division protein kinase 2; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 2; EC 2.7.11.22; EC 2.7.11.23; kinase Cdc2; MPF; p33 protein kinase; p33(CDK2); p33CDK2; p34 protein kinase | |
| Full length human recombinant protein of human CDK2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction