missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK20 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98621AF647
This item is not returnable.
View return policy
Description
CDK20 Polyclonal antibody specifically detects CDK20 in Human samples. It is validated for Western Blot
Specifications
| CDK20 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| CAK-kinase p42, CCRKEC 2.7.11.22, CDCHP42, CDK-activating kinase p42, cell cycle related kinase, Cell cycle-related kinase, Cell division protein kinase 20, cyclin-dependent kinase 20, Cyclin-dependent protein kinase H, Cyclin-kinase-activating kinase p42, EC 2.7.11, p42, PNQALRE | |
| Produced in rabbits immunized with E. coli-derived Human CDK20 fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 23552 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur