missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK2AP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566098
This item is not returnable.
View return policy
Description
Immunogen sequence: ATSSQYRQLLS DYGPPSLGYT QGTGNSQVPQ SKYAELLAII EELGKEI Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The protein encoded by this gene is a cyclin-dependent kinase 2 (CDK2) -associated protein which is thought to negatively regulate CDK2 activity by sequestering monomeric CDK2, and targeting CDK2 for proteolysis. This protein was found to also interact with DNA polymerase alpha/primase and mediate the phosphorylation of the large p180 subunit, which suggests a regulatory role in DNA replication during the S-phase of the cell cycle. This protein also forms a core subunit of the nucleosome remodeling and histone deacetylation (NURD) complex that epigenetically regulates embryonic stem cell differentiation. This gene thus plays a role in both cell-cycle and epigenetic regulation. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
| CDK2AP1 | |
| Polyclonal | |
| Unconjugated | |
| CDK2AP1 | |
| Apc10; CDK2 (cyclin-dependent kinase 2)-associated protein 1; Cdk2ap1; CDK2-associated protein 1; CDKAP1; cyclin dependent kinase 2 associated protein 1; cyclin-dependent kinase 2 associated protein 1; cyclin-dependent kinase 2-associated protein 1; Deleted in oral cancer 1; Deleted in oral cancer-1; Doc1; doc-1; DORC1; p12; p12DOC-1; Putative oral cancer suppressor; ST19 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8099 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14519 | |
| CDK2AP1 | |
| Recombinant Human CDK2AP1. Recombinant protein control fragment (Product #RP-106793). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction